Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019112.3 | NP_061985.2 | 12 | 1660 | ||
XM_006722616.1 | XP_006722679.1 | 12 | 1647 | |||
XM_006722617.2 | XP_006722680.1 | 12 | 1647 | |||
XM_011527628.2 | XP_011525930.1 | 12 | 1521 | |||
XM_011527629.1 | XP_011525931.1 | 12 | 1647 | |||
XM_011527630.1 | XP_011525932.1 | 12 | 1647 | |||
XM_011527631.1 | XP_011525933.1 | 12 | 1647 | |||
XM_011527632.1 | XP_011525934.1 | 8 | 1099 | |||
XM_011527633.2 | XP_011525935.1 | 12 | 1647 | |||
XM_011527634.1 | XP_011525936.1 | 12 | 1647 | |||
XM_011527635.1 | XP_011525937.1 | 12 | 1647 | |||
XM_017026142.1 | XP_016881631.1 | 12 | 1647 | |||
XM_017026143.1 | XP_016881632.1 | 12 | 1647 | |||
GenBank mRNA | AB055390.1 | - | 6 | 1015 | ||
AF250238.1 | - | 11 | 1429 | |||
AF328787.1 | - | 11 | 1545 |