Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020208.3 | NP_064593.1 | 11 | 1935 | ||
NM_022405.3 | NP_071800.1 | 10 | 1824 | |||
XM_011533847.2 | XP_011532149.1 | 10 | 2249 | |||
GenBank mRNA | AJ276207.1 | - | 11 | 1862 | ||
AJ276208.1 | - | 10 | 1751 | |||
AL389979.1 | - | 6 | 1999 | |||
BC126197.1 | - | 11 | 1875 | 8991913 | ||
BC136431.1 | - | 11 | 1875 | 9020420 |