Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021095.2 | NP_066918.2 | 17 | 2760 | ||
XM_006712128.1 | XP_006712191.1 | 17 | 2555 | |||
XM_006712129.1 | XP_006712192.1 | 17 | 2603 | |||
XM_006712130.1 | XP_006712193.1 | 17 | 2594 | |||
XM_011533146.2 | XP_011531448.1 | 15 | 2320 | |||
XM_017005216.1 | XP_016860705.1 | 14 | 2330 | |||
GenBank mRNA | AF069307.1 | - | 17 | 2656 | ||
AF081571.1 | - | 16 | 2472 | |||
AK075078.1 | - | 8 | 1198 | |||
AL096737.1 | - | 3 | 1814 | |||
BC012806.2 | - | 15 | 2322 | 3507776 | ||
BC015631.2 | - | 16 | 2473 | 3628126 |