Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC8A1 SIRNA Assays ›
Gene Symbol:
SLC8A1
Gene Name:
solute carrier family 8 member A1
Gene Aliases:
NCX1
Chromosome Location:
Chr.2: 40112146 - 40512452 on Build GRCh38
UniGene ID:
Hs.31961
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
117631
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC8A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112800.1
NP_001106271.1
1
1116
NM_001112801.1
NP_001106272.1
1
1116
NM_001112802.1
NP_001106273.1
2
1203
NM_001252624.1
NP_001239553.1
1
1116
NM_021097.2
NP_066920.1
1
1116
XM_005264514.3
XP_005264571.1
2
1298
XM_006712081.2
XP_006712144.1
2
1298
XM_006712082.3
XP_006712145.1
3
1400
XM_006712083.3
XP_006712146.1
2
1217
XM_006712084.3
XP_006712147.1
3
1498
XM_006712085.3
XP_006712148.1
2
2120
XM_011533050.1
XP_011531352.1
1
1113
XM_011533054.2
XP_011531356.1
2
1134
XM_011533055.2
XP_011531357.1
2
1261
XM_011533056.1
XP_011531358.1
2
1134
XM_011533057.2
XP_011531359.2
2
1282
XM_011533058.2
XP_011531360.1
2
1133
XM_017004745.1
XP_016860234.1
2
1282
XM_017004746.1
XP_016860235.1
2
1135
XM_017004747.1
XP_016860236.1
2
1337
XM_017004748.1
XP_016860237.1
2
1203
XM_017004749.1
XP_016860238.1
3
1403
XM_017004750.1
XP_016860239.1
4
1637
XM_017004751.1
XP_016860240.1
4
1634
XM_017004752.1
XP_016860241.1
3
1438
XM_017004753.1
XP_016860242.1
2
1134
XM_017004754.1
XP_016860243.1
2
1262
XM_017004755.1
XP_016860244.1
2
1261
XM_017004756.1
XP_016860245.1
2
1341
XM_017004757.1
XP_016860246.1
2
1336
XM_017004758.1
XP_016860247.1
2
1318
XM_017004759.1
XP_016860248.1
2
1324
XM_017004760.1
XP_016860249.1
2
1298
XM_017004761.1
XP_016860250.1
2
1341
XM_017004762.1
XP_016860251.1
2
1316
XM_017004763.1
XP_016860252.1
2
1298
XM_017004764.1
XP_016860253.1
2
1133
GenBank mRNA
AF108388.1
-
1
1092
AF108389.1
-
1
1092
AF128524.1
-
1
1114
AK291696.1
-
2
1203
BX537482.1
-
2
1202
BX648299.1
-
2
1283
M91368.1
-
1172
M96368.1
-
1
1133
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
regulation of the force of heart contraction
regulation of heart rate
regulation of sodium ion transport
ion transport
cellular sodium ion homeostasis
muscle contraction
response to nutrient
response to glucose
regulation of cell communication by electrical coupling
positive regulation of fibroblast migration
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of cardiac muscle contraction by calcium ion signaling
vascular smooth muscle contraction
telencephalon development
positive regulation of bone mineralization
response to ATP
cellular response to reactive oxygen species
sodium ion transmembrane transport
response to immobilization stress
response to muscle stretch
response to drug
response to hydrogen peroxide
relaxation of smooth muscle
negative regulation of cytosolic calcium ion concentration
cardiac muscle cell development
calcium ion homeostasis
relaxation of cardiac muscle
cardiac muscle contraction
cytosolic calcium ion transport
calcium ion transport into cytosol
calcium ion import
calcium ion transmembrane transport
cellular response to caffeine
cellular response to cAMP
sodium ion export
cellular response to hypoxia
membrane depolarization during cardiac muscle cell action potential
cell communication by electrical coupling involved in cardiac conduction
sodium ion import
positive regulation of the force of heart contraction
calcium ion export
regulation of cardiac conduction
calcium:sodium antiporter activity
calcium ion binding
protein binding
calmodulin binding
cytoskeletal protein binding
ankyrin binding
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-zpscg:80/10.239.160.225:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0