Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243137.1 | NP_001230066.1 | 2, 3 | 238 | ||
NM_002605.2 | NP_002596.1 | 2, 3 | 440 | |||
NM_173454.1 | NP_775656.1 | 2, 3 | 440 | |||
XM_017022308.1 | XP_016877797.1 | 2, 3 | 977 | |||
XM_017022309.1 | XP_016877798.1 | 2, 3 | 150 | |||
XM_017022310.1 | XP_016877799.1 | 2, 3 | 977 | |||
XM_017022311.1 | XP_016877800.1 | 2, 3 | 977 | |||
GenBank mRNA | AF332653.1 | - | 2, 3 | 387 | ||
AF388183.1 | - | 2, 3 | 433 | |||
AF388184.1 | - | 2, 3 | 433 | |||
AF388185.1 | - | 2, 3 | 433 | |||
AF388186.1 | - | 2, 3 | 433 | |||
AF388187.1 | - | 2, 3 | 433 | |||
AK074280.1 | - | 2, 3 | 238 | |||
AK127232.1 | - | 1, 2 | 1938 | |||
BC060762.1 | - | 3, 4 | 503 | 30344110 | ||
BC075822.1 | - | 2, 3 | 331 | 6296489 |