Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184779.1 | NP_001171708.1 | 13, 14 | 1855 | ||
NM_001184780.1 | NP_001171709.1 | 13, 14 | 1982 | |||
NM_024505.3 | NP_078781.3 | 13, 14 | 1939 | |||
NR_033671.2 | 14, 15 | 2170 | ||||
NR_033672.1 | 14, 15 | 2086 | ||||
GenBank mRNA | AF317889.1 | - | 10, 11 | 1370 | ||
AF325189.1 | - | 14, 15 | 2061 | |||
AF325190.1 | - | 14, 15 | 2145 | |||
AF353088.1 | - | 13, 14 | 1834 | |||
AF353089.1 | - | 13, 14 | 1918 | |||
AK026011.1 | - | 10, 11 | 1424 | |||
AK074058.1 | - | 11, 12 | 1608 | |||
AK074071.1 | - | 12, 13 | 5057 | |||
AK314689.1 | - | 13, 14 | 1918 | |||
AY166601.1 | - | 9, 10 | 1165 | |||
BC125097.1 | - | 13, 14 | 1848 | 40118286 | ||
BC125098.1 | - | 14, 15 | 1952 | 40118288 | ||
DQ413001.1 | - | 10, 11 | 1298 |