Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024854.3 | NP_079130.2 | 4 | 447 | ||
XM_006719153.3 | XP_006719216.1 | 4 | 421 | |||
GenBank mRNA | AK025681.1 | - | 4 | 349 | ||
AK125461.1 | - | 4 | 860 | |||
AK293820.1 | - | 4 | 436 | |||
AK307710.1 | - | 4 | 346 | |||
AL832441.1 | - | 4 | 374 | |||
BC021662.1 | - | 4 | 348 | 3826938 | ||
DB041552.1 | - | 4 | 447 |