Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025059.3 | NP_079335.2 | 2 | 248 | ||
XM_011536147.2 | XP_011534449.1 | 2 | 286 | |||
XM_011536148.2 | XP_011534450.1 | 2 | 289 | |||
GenBank mRNA | AK026958.1 | - | 2 | 161 | ||
AK225330.1 | - | 2 | 162 | |||
BC060803.1 | - | 2 | 197 | 5298914 | ||
BI562667.1 | - | 2 | 202 | |||
DB232703.1 | - | 2 | 248 |