Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004434.2 | NP_001004434.1 | 6 | 1053 | ||
NM_032513.4 | NP_115902.1 | 5 | 906 | |||
GenBank mRNA | AF370409.1 | - | 6 | 900 | ||
AK023491.1 | - | 1 | 75 | |||
BC006251.2 | - | 5 | 881 | 3948165 |