Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX4I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032609.2 | NP_115998.2 | 4, 5 | 452 | ||
XM_005260579.4 | XP_005260636.1 | 3, 4 | 454 | |||
GenBank mRNA | AF257180.1 | - | 4, 5 | 452 | ||
BC057779.1 | - | 4, 5 | 428 | 30337282 | ||
HE647859.1 | - | 3, 4 | 377 |