Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080283.3 | NP_525022.2 | 4, 5 | 620 | ||
XM_017024011.1 | XP_016879500.1 | 4, 5 | 585 | |||
XM_017024012.1 | XP_016879501.1 | 4, 5 | 689 | |||
XM_017024013.1 | XP_016879502.1 | 4, 5 | 509 | |||
XM_017024014.1 | XP_016879503.1 | 4, 5 | 1109 | |||
GenBank mRNA | AF423307.1 | - | 3, 4 | 477 | ||
AK057068.1 | - | 4, 5 | 552 | |||
AY028899.1 | - | 4, 5 | 620 | |||
BC062472.1 | - | 4, 5 | 531 | |||
CR627382.1 | - | 3, 4 | 426 |