Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080283.3 | NP_525022.2 | 18 | 2509 | ||
XM_017024011.1 | XP_016879500.1 | 18 | 2474 | |||
XM_017024012.1 | XP_016879501.1 | 18 | 2578 | |||
XM_017024013.1 | XP_016879502.1 | 18 | 2398 | |||
XM_017024014.1 | XP_016879503.1 | 18 | 2998 | |||
XM_017024015.1 | XP_016879504.1 | 15 | 2021 | |||
GenBank mRNA | AF423307.1 | - | 17 | 2366 | ||
AK057068.1 | - | 18 | 2441 | |||
AK074491.1 | - | 11 | 1318 | |||
AY028899.1 | - | 18 | 2509 | |||
CR627382.1 | - | 17 | 2315 |