Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021094.3 | NP_066580.1 | 4 | 669 | ||
NM_134431.3 | NP_602307.1 | 6 | 1139 | |||
XM_005253474.3 | XP_005253531.1 | 5 | 560 | |||
XM_005253477.3 | XP_005253534.1 | 4 | 447 | |||
XM_011520818.1 | XP_011519120.1 | 5 | 567 | |||
XM_011520819.1 | XP_011519121.1 | 5 | 714 | |||
XM_011520820.2 | XP_011519122.1 | 5 | 2036 | |||
XM_017019849.1 | XP_016875338.1 | 5 | 567 | |||
XM_017019850.1 | XP_016875339.1 | 4 | 447 | |||
GenBank mRNA | AF085224.1 | - | 5 | 628 | ||
AF279784.1 | - | 6 | 1139 | |||
AK296044.1 | - | 3 | 280 | |||
AK316249.1 | - | 4 | 699 | |||
BC042452.1 | - | 4 | 449 | 4820050 | ||
U21943.1 | - | 4 | 471 |