Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC23A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144889.1 | NP_001138361.1 | 1 | 230 | ||
NM_001144890.1 | NP_001138362.1 | 1 | 230 | |||
NM_144712.4 | NP_653313.3 | 1 | 230 | |||
GenBank mRNA | AK055730.1 | - | 1 | 192 | ||
AK298256.1 | - | 1 | 140 | |||
AK298277.1 | - | 1 | 139 | |||
BC030243.1 | - | 1 | 128 | 5188653 | ||
DC361649.1 | - | 1 | 233 |