Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC23A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144889.1 | NP_001138361.1 | 12 | 1686 | ||
NM_001144890.1 | NP_001138362.1 | 12 | 1710 | |||
NM_144712.4 | NP_653313.3 | 11 | 1335 | |||
GenBank mRNA | AK298256.1 | - | 12 | 1596 | ||
AK298277.1 | - | 12 | 1619 | |||
BC030243.1 | - | 11 | 1233 | 5188653 |