Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145176.2 | NP_660159.1 | 2 | 504 | ||
XM_006715349.3 | XP_006715412.1 | 2 | 521 | |||
XM_017010311.1 | XP_016865800.1 | 2 | 521 | |||
GenBank mRNA | AK122628.1 | - | 2 | 496 | ||
AK226098.1 | - | 2 | 375 | |||
AL833602.1 | - | 2 | 488 | |||
AY046419.1 | - | 2 | 466 | |||
BC070149.1 | - | 2 | 438 | 30371087 | ||
DB253903.1 | - | 2 | 507 |