Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042450.2 | NP_001035915.1 | 15 | 1987 | ||
NM_001270648.1 | NP_001257577.1 | 14 | 1906 | |||
NM_001270649.1 | NP_001257578.1 | 14 | 1879 | |||
NM_001282417.1 | NP_001269346.1 | 15 | 2096 | |||
NM_152351.4 | NP_689564.3 | 15 | 2035 | |||
XM_011523652.1 | XP_011521954.1 | 16 | 2254 | |||
XM_011523653.2 | XP_011521955.1 | 15 | 2226 | |||
XM_017024191.1 | XP_016879680.1 | 16 | 2303 | |||
GenBank mRNA | AK057946.1 | - | 15 | 2036 | ||
AK298345.1 | - | 14 | 1865 | |||
BC034380.1 | - | 15 | 1932 | |||
BC039868.1 | - | 15 | 2113 | 4157703 | ||
BC062617.1 | - | 14 | 1860 |