Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152679.3 | NP_689892.1 | 3 | 1403 | ||
GenBank mRNA | AK126542.1 | - | 3 | 1403 | ||
AY704414.1 | - | 3 | 1184 | |||
BC012048.1 | - | 3 | 1261 | 3502817 | ||
BC019066.2 | - | 3 | 1314 | 4897519 |