Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136506.2 | NP_001129978.2 | 4 | 1138 | ||
NM_173586.2 | NP_775857.2 | 4 | 1138 | |||
GenBank mRNA | BC034394.1 | - | 4 | 1138 | 5189722 |