Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002442.3 | NP_002433.1 | 15 | 1534 | ||
XM_006719403.3 | XP_006719466.1 | 15 | 1495 | |||
XM_006719404.3 | XP_006719467.1 | 10 | 1313 | |||
XM_011538362.2 | XP_011536664.1 | 14 | 1471 | |||
XM_011538363.2 | XP_011536665.1 | 14 | 1438 | |||
XM_011538364.2 | XP_011536666.1 | 11 | 2078 | |||
XM_011538365.2 | XP_011536667.1 | 12 | 1197 | |||
XM_011538366.2 | XP_011536668.1 | 12 | 2205 | |||
XM_011538368.2 | XP_011536670.1 | 10 | 1153 | |||
XM_011538370.2 | XP_011536672.1 | 10 | 1079 | |||
XM_011538371.2 | XP_011536673.1 | 10 | 1105 | |||
GenBank mRNA | AB012851.1 | - | 15 | 1509 | ||
AK023262.1 | - | 8 | 954 |