Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172811.1 | NP_001166282.1 | 10 | 1203 | ||
NM_001172813.1 | NP_001166284.1 | 11 | 1448 | |||
NM_001172814.1 | NP_001166285.1 | 9 | 1290 | |||
NM_001172815.2 | NP_001166286.1 | 11 | 1317 | |||
NM_173851.2 | NP_776250.2 | 8 | 1260 | |||
GenBank mRNA | AK298294.1 | - | 12 | 1451 | ||
AL713790.1 | - | 11 | 1289 | |||
AY117411.1 | - | 8 | 1030 | |||
BC126446.1 | - | 9 | 1129 | |||
BC143953.1 | - | 9 | 1130 | |||
EF560713.1 | - | 10 | 1203 |