Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO4C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180991.4 | NP_851322.3 | 1, 2 | 650 | ||
XM_011543372.1 | XP_011541674.1 | 3, 4 | 301 | |||
GenBank mRNA | AF401643.1 | - | 1, 2 | 363 | ||
AK128854.1 | - | 1, 2 | 378 | |||
AY273896.2 | - | 1, 2 | 650 | |||
BX647349.1 | - | 1, 2 | 543 |