Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205206.1 | NP_001192135.1 | 3 | 712 | ||
NM_001205207.1 | NP_001192136.1 | 2 | 461 | |||
NM_194255.2 | NP_919231.1 | 3 | 712 | |||
XM_005261164.3 | XP_005261221.1 | 3 | 685 | |||
XM_011529696.2 | XP_011527998.1 | 4 | 1144 | |||
XM_011529698.2 | XP_011528000.1 | 3 | 748 | |||
XM_011529700.2 | XP_011528002.1 | 3 | 1007 | |||
XM_011529701.2 | XP_011528003.1 | 4 | 957 | |||
XM_011529702.2 | XP_011528004.1 | 3 | 671 | |||
XM_011529703.2 | XP_011528005.1 | 3 | 739 | |||
XM_011529704.2 | XP_011528006.1 | 3 | 655 | |||
XM_011529706.2 | XP_011528008.1 | 2 | 1100 | |||
XM_011529709.2 | XP_011528011.1 | 3 | 926 | |||
XM_017028443.1 | XP_016883932.1 | 3 | 836 | |||
XM_017028444.1 | XP_016883933.1 | 4 | 1150 | |||
XM_017028445.1 | XP_016883934.1 | 4 | 1720 | |||
XM_017028446.1 | XP_016883935.1 | 3 | 1010 | |||
GenBank mRNA | AF004354.1 | - | 2 | 630 | ||
AK303168.1 | - | 2 | 448 | |||
AK307623.1 | - | 3 | 716 | |||
AK310797.1 | - | 3 | 880 | |||
AK313125.1 | - | 3 | 676 | |||
BC003068.2 | - | 3 | 657 | 3509422 | ||
DC311922.1 | - | 2 | 461 | |||
S78996.1 | - | 2 | 559 | |||
U15939.1 | - | 3 | 653 | |||
U17566.1 | - | 3 | 653 | |||
U19720.1 | - | 3 | 654 |