Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205207.1 | NP_001192136.1 | 5 | 2187 | ||
NM_194255.2 | NP_919231.1 | 6 | 2438 | |||
XM_005261164.3 | XP_005261221.1 | 6 | 2411 | |||
XM_011529696.2 | XP_011527998.1 | 7 | 2870 | |||
XM_011529698.2 | XP_011528000.1 | 6 | 2474 | |||
XM_011529700.2 | XP_011528002.1 | 6 | 2733 | |||
XM_011529701.2 | XP_011528003.1 | 7 | 2683 | |||
XM_011529702.2 | XP_011528004.1 | 6 | 2397 | |||
XM_011529703.2 | XP_011528005.1 | 6 | 2465 | |||
XM_011529704.2 | XP_011528006.1 | 6 | 2381 | |||
XM_011529706.2 | XP_011528008.1 | 5 | 2826 | |||
XM_011529709.2 | XP_011528011.1 | 6 | 2652 | |||
XM_017028443.1 | XP_016883932.1 | 6 | 2562 | |||
GenBank mRNA | AF004354.1 | - | 5 | 2356 | ||
BC128247.1 | - | 1 | 906 | |||
U17566.1 | - | 6 | 2378 | |||
U19720.1 | - | 6 | 2380 |