Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004245.3 | NP_004236.1 | 9 | 1159 | ||
NM_201631.3 | NP_963925.2 | 10 | 1405 | |||
XM_011522230.2 | XP_011520532.1 | 4 | 587 | |||
XM_017022729.1 | XP_016878218.1 | 4 | 611 | |||
GenBank mRNA | AF035960.1 | - | 10 | 1405 | ||
AF035961.1 | - | 9 | 1159 | |||
AK298479.1 | - | 6 | 821 | |||
BC119009.1 | - | 10 | 1405 | 40085374 | ||
BC122859.1 | - | 10 | 1405 | 40085372 |