Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000178.3 | NP_000169.1 | 3 | 326 | ||
NM_001322494.1 | NP_001309423.1 | 3 | 393 | |||
NM_001322495.1 | NP_001309424.1 | 3 | 1155 | |||
GenBank mRNA | AB459500.1 | - | 2 | 227 | ||
AK293144.1 | - | 3 | 217 | |||
AK298292.1 | - | 2 | 201 | |||
AK311421.1 | - | 3 | 297 | |||
AK311567.1 | - | 2 | 108 | |||
AK312492.1 | - | 3 | 306 | |||
BC007927.1 | - | 3 | 313 | 3946317 | ||
DC328543.1 | - | 3 | 393 | |||
EU668322.1 | - | 3 | 313 | |||
EU794618.1 | - | 3 | 313 | |||
KU178105.1 | - | 2 | 227 | |||
KU178106.1 | - | 2 | 227 | |||
U34683.1 | - | 3 | 267 |