Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000178.3 | NP_000169.1 | 13 | 1616 | ||
NM_001322494.1 | NP_001309423.1 | 13 | 1683 | |||
NM_001322495.1 | NP_001309424.1 | 13 | 2445 | |||
GenBank mRNA | BC007927.1 | - | 13 | 1603 | 3946317 | |
EU668322.1 | - | 13 | 1603 | |||
EU794618.1 | - | 13 | 1603 | |||
U34683.1 | - | 13 | 1557 |