Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000411.6 | NP_000402.3 | 12 | 5958 | ||
NM_001242784.1 | NP_001229713.1 | 12 | 6892 | |||
NM_001242785.1 | NP_001229714.1 | 12 | 6051 | |||
GenBank mRNA | BI094308.1 | - | 1 | 511 |