Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293312.1 | NP_001280241.1 | 9 | 1092 | ||
NM_001349.3 | NP_001340.2 | 10 | 1150 | |||
XM_017003489.1 | XP_016858978.1 | 10 | 1121 | |||
GenBank mRNA | AK129521.1 | - | 10 | 1092 | ||
AK222476.1 | - | 10 | 980 | |||
AK290607.1 | - | 10 | 1007 | |||
BC000629.2 | - | 10 | 983 | 3344322 | ||
BC107749.1 | - | 10 | 963 | 6709590 | ||
BT006710.1 | - | 10 | 934 | |||
CR749809.1 | - | 9 | 900 | |||
J05032.1 | - | 10 | 1024 |