Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258040.2 | NP_001244969.1 | 3 | 647 | ||
NM_001258041.2 | NP_001244970.1 | 4 | 767 | |||
NM_001258042.2 | NP_001244971.1 | 3 | 647 | |||
NM_001289094.1 | NP_001276023.1 | 4 | 680 | |||
NM_002109.5 | NP_002100.2 | 4 | 767 | |||
GenBank mRNA | AK000498.1 | - | 4 | 447 | ||
AK055917.1 | - | 3 | 261 | |||
AK124831.1 | - | 3 | 1957 | |||
AK225776.1 | - | 4 | 417 | |||
AK295219.1 | - | 3 | 303 | |||
AK302295.1 | - | 3 | 261 | |||
AK307669.1 | - | 4 | 360 | |||
AY995220.1 | - | 4 | 344 | |||
BC011807.2 | - | 4 | 374 | 4331277 | ||
BC080514.1 | - | 4 | 372 | 2821065 | ||
X05345.1 | - | 4 | 421 | |||
Z11518.1 | - | 4 | 902 |