Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002887.3 | NP_002878.2 | 8 | 952 | ||
GenBank mRNA | AK222797.1 | - | 8 | 928 | ||
AK302161.1 | - | 7 | 810 | |||
AK314795.1 | - | 8 | 940 | |||
BC000528.2 | - | 8 | 930 | 2961565 | ||
BC014619.2 | - | 8 | 930 | 2961565 | ||
BT007394.1 | - | 8 | 911 | |||
S80343.1 | - | 8 | 938 |