Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002471.3 | NP_002462.2 | 25 | 3334 | ||
GenBank mRNA | BC117511.1 | - | 2 | 63 | ||
BC132667.1 | - | 24 | 3293 | 40146689 | ||
D00943.1 | - | 23 | 3263 |