Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306079.1 | NP_001293008.1 | 4 | 581 | ||
NM_004341.4 | NP_004332.2 | 4 | 581 | |||
XM_005264555.3 | XP_005264612.1 | 4 | 714 | |||
XM_006712101.2 | XP_006712164.1 | 4 | 714 | |||
GenBank mRNA | AB621827.1 | - | 4 | 499 | ||
AK226105.1 | - | 4 | 563 | |||
BC065510.1 | - | 4 | 581 | 5551082 | ||
D78586.1 | - | 4 | 445 |