Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004990.3 | NP_004981.2 | 4 | 526 | ||
XM_006719398.3 | XP_006719461.1 | 4 | 437 | |||
GenBank mRNA | AK092986.1 | - | 3 | 446 | ||
AK122956.1 | - | 5 | 1074 | |||
AK222767.1 | - | 4 | 408 | |||
AK290857.1 | - | 4 | 397 | |||
AK293945.1 | - | 4 | 413 | |||
BC002384.2 | - | 4 | 394 | 2961615 | ||
BC006328.2 | - | 4 | 394 | 4139330 | ||
BC011639.2 | - | 4 | 396 | 4120680 | ||
BC011849.2 | - | 4 | 394 | 4135154 | ||
BC015011.1 | - | 4 | 386 | |||
BT007338.1 | - | 4 | 388 | |||
D84224.1 | - | 4 | 411 | |||
DA489707.1 | - | 4 | 527 | |||
X94754.1 | - | 4 | 395 |