Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002477.1 | NP_002468.1 | 5 | 465 | ||
XM_006713886.2 | XP_006713949.1 | 5 | 842 | |||
XM_017008245.1 | XP_016863734.1 | 6 | 2143 | |||
XM_017008246.1 | XP_016863735.1 | 7 | 2012 | |||
XM_017008247.1 | XP_016863736.1 | 5 | 1602 | |||
XM_017008248.1 | XP_016863737.1 | 6 | 625 | |||
GenBank mRNA | AK123974.1 | - | 6 | 2761 | ||
AK125529.1 | - | 6 | 1585 | |||
L03785.1 | - | 5 | 465 |