Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145933.1 | NP_001139405.1 | 7 | 1126 | ||
NM_001145934.1 | NP_001139406.1 | 6 | 1071 | |||
NM_012253.3 | NP_036385.3 | 7 | 1144 | |||
GenBank mRNA | AK292224.1 | - | 7 | 1016 | ||
AK292261.1 | - | 7 | 1020 | |||
AK301724.1 | - | 7 | 945 | |||
AK302044.1 | - | 7 | 1002 | |||
AK302261.1 | - | 3 | 409 | |||
BC025382.2 | - | 7 | 1020 | 4825931 | ||
X91817.1 | - | 6 | 910 |