Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POLL SIRNA Assays ›
Gene Symbol:
POLL
Gene Name:
polymerase (DNA) lambda
Gene Aliases:
BETAN, POLKAPPA
Chromosome Location:
Chr.10: 101578882 - 101588270 on Build GRCh38
UniGene ID:
Hs.523230
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
117948
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POLL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174084.1
NP_001167555.1
6
1475
NM_001174085.1
NP_001167556.1
6
1136
NM_001308382.1
NP_001295311.1
3
317
NM_013274.3
NP_037406.1
6
1412
NR_033406.1
6
1011
XM_006717775.1
XP_006717838.1
3
1080
XM_006717776.1
XP_006717839.1
4
308
XM_006717777.1
XP_006717840.1
2
1038
XM_011539650.1
XP_011537952.1
6
1080
XM_011539651.1
XP_011537953.1
6
1080
XM_011539652.1
XP_011537954.1
6
1368
XM_011539653.1
XP_011537955.1
6
1361
XM_011539654.1
XP_011537956.1
6
804
XM_011539655.1
XP_011537957.1
6
804
XM_011539656.1
XP_011537958.1
5
762
XM_011539657.1
XP_011537959.1
6
1307
XM_011539659.1
XP_011537961.1
3
1356
XM_011539660.1
XP_011537962.1
4
612
XM_011539662.1
XP_011537964.1
3
1080
XM_011539663.1
XP_011537965.1
4
308
XM_011539664.1
XP_011537966.1
2
1038
XM_011539665.2
XP_011537967.1
3
626
XM_011539666.1
XP_011537968.1
3
304
XM_011539667.1
XP_011537969.1
5
603
XM_017016086.1
XP_016871575.1
4
1470
XM_017016089.1
XP_016871578.1
4
885
XM_017016091.1
XP_016871580.1
3
1356
XM_017016093.1
XP_016871582.1
3
563
GenBank mRNA
AF161019.1
-
5
1041
AF283478.1
-
5
980
AJ131890.1
-
6
1351
AK021600.1
-
6
1083
AK022476.1
-
6
989
AK127896.1
-
1
747
AK292225.1
-
6
1058
AK293421.1
-
5
633
AK293602.1
-
4
308
AK293657.1
-
4
754
AK300862.1
-
3
296
AY302442.1
-
5
704
BC003548.1
-
2
970
3536366
BC068529.1
-
6
1364
5267859
BI906881.1
-
3
277
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-directed DNA polymerase
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
nucleotide-excision repair
double-strand break repair via nonhomologous end joining
somatic hypermutation of immunoglobulin genes
DNA biosynthetic process
DNA binding
DNA-directed DNA polymerase activity
lyase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline