Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTDSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014754.2 | NP_055569.1 | 3 | 458 | ||
GenBank mRNA | AK293036.1 | - | 3 | 442 | ||
BC004192.2 | - | 3 | 408 | 3509418 | ||
BC004390.1 | - | 3 | 408 | 3634879 | ||
D14694.1 | - | 3 | 405 |