Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017582.6 | NP_060052.3 | 3 | 564 | ||
GenBank mRNA | AK293986.1 | - | 3 | 187 | ||
AY112698.1 | - | 3 | 539 | |||
AY948200.1 | - | 3 | 547 | |||
BC070158.1 | - | 2 | 89 | |||
BC107057.2 | - | 4 | 232 | |||
BC148185.2 | - | 3 | 241 | |||
BQ278712.1 | - | 564 |