Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136035.2 | NP_001129507.1 | 4 | 459 | ||
NM_001142554.1 | NP_001136026.1 | 3 | 580 | |||
NM_017722.3 | NP_060192.1 | 3 | 580 | |||
XM_011528124.1 | XP_011526426.1 | 4 | 378 | |||
XM_011528125.1 | XP_011526427.1 | 5 | 911 | |||
XM_017026944.1 | XP_016882433.1 | 5 | 902 | |||
XM_017026945.1 | XP_016882434.1 | 5 | 902 | |||
XM_017026947.1 | XP_016882436.1 | 5 | 901 | |||
GenBank mRNA | AF196479.1 | - | 3 | 330 | ||
AK000251.1 | - | 4 | 418 | |||
AK292377.1 | - | 4 | 459 | |||
AK295245.1 | - | 4 | 359 | |||
AK304636.1 | - | 4 | 405 | |||
BC002492.2 | - | 3 | 410 | 3051390 | ||
BC018302.1 | - | 3 | 575 | 3463518 | ||
BC040126.1 | - | 3 | 449 | 5730044 |