Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257158.1 | NP_001244087.1 | 7, 8 | 826 | ||
NM_001257159.1 | NP_001244088.1 | 6, 7 | 778 | |||
NM_018718.2 | NP_061188.1 | 7, 8 | 826 | |||
NR_046443.1 | 7, 8 | 750 | ||||
XM_011516709.2 | XP_011515011.1 | 8, 9 | 787 | |||
XM_011516710.2 | XP_011515012.1 | 6, 7 | 602 | |||
GenBank mRNA | AF429308.1 | - | 7, 8 | 672 | ||
AJ278890.1 | - | 7, 8 | 629 | |||
AK026098.1 | - | 7, 8 | 626 | |||
AK298618.1 | - | 6, 7 | 594 | |||
AK311382.1 | - | 7, 8 | 567 | |||
AK314676.1 | - | 7, 8 | 605 | |||
AL359617.1 | - | 1, 2 | 127 | |||
BC018789.1 | - | 7, 8 | 608 | |||
BC056162.1 | - | 7, 8 | 599 | 6142659 |