Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AS3MT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020682.3 | NP_065733.2 | 7 | 714 | ||
GenBank mRNA | AF226730.1 | - | 7 | 651 | ||
AK057833.1 | - | 7 | 733 | |||
AK225972.1 | - | 7 | 733 | |||
AK309737.1 | - | 7 | 646 | |||
BC119637.1 | - | 7 | 618 | |||
BC119638.2 | - | 7 | 618 |
Gene Symbol: BORCS7-ASMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037644.1 | 11 | 979 |