Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242828.1 | NP_001229757.1 | 5 | 716 | ||
NM_001242830.1 | NP_001229759.1 | 4 | 635 | |||
NM_001301856.1 | NP_001288785.1 | 4 | 868 | |||
NM_021814.4 | NP_068586.1 | 4 | 635 | |||
GenBank mRNA | AB209798.1 | - | 4 | 383 | ||
AF231981.1 | - | 3 | 264 | |||
AF338241.1 | - | 4 | 608 | |||
AK074889.1 | - | 3 | 269 | |||
AK125098.1 | - | 4 | 712 | |||
AK299674.1 | - | 4 | 376 | |||
AK302948.1 | - | 5 | 487 | |||
AL136939.1 | - | 4 | 383 | |||
BC067123.1 | - | 4 | 384 | 6160926 |