Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242828.1 | NP_001229757.1 | 9 | 2604 | ||
NM_001242830.1 | NP_001229759.1 | 7 | 2398 | |||
NM_001301856.1 | NP_001288785.1 | 8 | 2756 | |||
NM_021814.4 | NP_068586.1 | 8 | 2523 | |||
GenBank mRNA | AB209798.1 | - | 7 | 2146 | ||
AF052129.1 | - | 1 | 712 | |||
AF111849.1 | - | 2 | 1543 | |||
AF338241.1 | - | 8 | 2496 | |||
AK074748.1 | - | 3 | 1620 | |||
AK074889.1 | - | 7 | 2157 | |||
AK125098.1 | - | 8 | 2601 | |||
AL136939.1 | - | 8 | 2270 | |||
BC017270.2 | - | 3 | 1567 | |||
BC067123.1 | - | 8 | 2272 | 6160926 |