Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022132.4 | NP_071415.1 | 4 | 492 | ||
XM_005248567.1 | XP_005248624.1 | 4 | 394 | |||
XM_011543529.2 | XP_011541831.1 | 4 | 394 | |||
XM_017009688.1 | XP_016865177.1 | 4 | 394 | |||
GenBank mRNA | AB050049.1 | - | 4 | 453 | ||
AF261884.1 | - | 4 | 427 | |||
AF301000.1 | - | 4 | 453 | |||
AF310971.1 | - | 4 | 474 | |||
AK310194.1 | - | 4 | 450 | |||
BC014897.1 | - | 4 | 492 | 3910739 | ||
BC065027.1 | - | 4 | 412 | 6043576 | ||
KU178741.1 | - | 4 | 354 | |||
KU178742.1 | - | 4 | 354 |