Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024560.2 | NP_078836.1 | 2 | 523 | ||
XM_005269150.2 | XP_005269207.1 | 2 | 457 | |||
XM_005269151.4 | XP_005269208.1 | 2 | 535 | |||
XM_011538740.2 | XP_011537042.1 | 1 | 121 | |||
GenBank mRNA | AK025616.1 | - | 2 | 474 | ||
AX764480.1 | - | 2 | 673 | |||
AX764482.1 | - | 2 | 429 | |||
BC009317.2 | - | 2 | 523 | 4120563 | ||
BC015769.1 | - | 2 | 523 | 4902942 |