Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193424.1 | NP_001180353.1 | 3 | 475 | ||
NM_001193425.1 | NP_001180354.1 | 3 | 433 | |||
NM_024670.3 | NP_078946.1 | 2 | 420 | |||
NR_034181.1 | 2 | 329 | ||||
XM_006717503.3 | XP_006717566.1 | 3 | 676 | |||
XM_011519662.2 | XP_011517964.1 | 3 | 464 | |||
XM_017016637.1 | XP_016872126.1 | 2 | 370 | |||
GenBank mRNA | AK027067.1 | - | 2 | 226 | ||
AK222565.1 | - | 2 | 236 | |||
AK225369.1 | - | 2 | 228 | |||
AL834488.1 | - | 1 | 84 | |||
BC007754.2 | - | 2 | 420 | 4109807 | ||
CR457372.1 | - | 1 | 189 | |||
DB040298.1 | - | 3 | 475 |