Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199957.1 | NP_001186886.1 | 4 | 587 | ||
NM_001199958.1 | NP_001186887.1 | 5 | 636 | |||
NM_002492.3 | NP_002483.1 | 6 | 743 | |||
GenBank mRNA | AF047181.1 | - | 6 | 720 | ||
BC005271.1 | - | 6 | 734 | 3997377 | ||
BC009796.1 | - | 6 | 738 | 4284776 | ||
BC017909.1 | - | 6 | 733 | |||
BC093071.1 | - | 4 | 564 | 30325282 | ||
BX648034.1 | - | 4 | 581 |