Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNKS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025235.3 | NP_079511.1 | 6, 7 | 1032 | ||
XM_005270185.4 | XP_005270242.2 | 6, 7 | 1063 | |||
XM_011540213.1 | XP_011538515.1 | 6, 7 | 1063 | |||
XM_017016696.1 | XP_016872185.1 | 6, 7 | 1000 | |||
XM_017016697.1 | XP_016872186.1 | 6, 7 | 1016 | |||
XM_017016698.1 | XP_016872187.1 | 6, 7 | 727 | |||
XM_017016699.1 | XP_016872188.1 | 6, 7 | 1016 | |||
XM_017016700.1 | XP_016872189.1 | 6, 7 | 613 | |||
XM_017016701.1 | XP_016872190.1 | 6, 7 | 1063 | |||
GenBank mRNA | AF264912.1 | - | 6, 7 | 735 | ||
AF305081.1 | - | 6, 7 | 1037 | |||
AF309033.1 | - | 6, 7 | 986 | |||
AF329696.1 | - | 6, 7 | 874 | |||
AF342982.1 | - | 6, 7 | 735 | |||
AF438201.1 | - | 6, 7 | 1114 | |||
AK314612.1 | - | 6, 7 | 1000 |