Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318051.1 | NP_001304980.1 | 4 | 413 | ||
NM_002495.3 | NP_002486.1 | 5 | 487 | |||
NR_134473.1 | 6 | 659 | ||||
NR_134474.1 | 6 | 576 | ||||
NR_134475.1 | 6 | 611 | ||||
GenBank mRNA | AF020351.1 | - | 5 | 465 | ||
AK307973.1 | - | 5 | 464 | |||
BC005270.1 | - | 5 | 481 | 3681312 | ||
BU145342.1 | - | 4 | 387 | |||
BU954271.1 | - | 5 | 479 | |||
CD692133.1 | - | 6 | 627 | |||
KU178226.1 | - | 5 | 457 | |||
KU178227.1 | - | 6 | 629 |